Merck
  • A novel molecularly imprinted polymer for simultaneous extraction and determination of sudan dyes by on-line solid phase extraction and high performance liquid chromatography.
A novel molecularly imprinted polymer for simultaneous extraction and determination of sudan dyes by on-line solid phase extraction and high performance liquid chromatography.
Journal of chromatography. A (2010-10-01)
Chuande Zhao, Ting Zhao, Xiaoyan Liu, Haixia Zhang
ABSTRACT
A novel molecularly imprinted polymer was synthesized with attapulgite employed as matrix, which is simple and time-saving. In this method, sudan I was chosen as template molecule, 2-vinylpyridine as functional monomer and ethylene glycol dimethacrylate as cross-linking agent, respectively. The imprinted polymer was characterized by the infrared spectroscopy and transmission electron microscopy. Then the selectivity experiments were performed on sudan dyes and the recognition coefficients for sudan I, sudan II, sudan III and sudan IV were 2.9, 1.9, 1.9 and 2.3, respectively. As the packing material of solid-phase extraction, the imprinted polymer has been applied to on-line concentration of the four sudan dyes in samples from Yellow River water, tomato sauce and sausage. The corresponding analytical methods to determine these sudan dyes have been developed. The limits of detection for these sudan dyes were in the range of 0.01-0.05 ng mL(-1) for Yellow River water, 1.0-3.0 ng g(-1) for tomato sauce and 0.8-3.0 ng g(-1) for sausage.
Sigma-Aldrich
Sudan I, Dye content ≥95 %
Pricing and availability is not currently available.
Supelco
Sudan I, analytical standard
Pricing and availability is not currently available.