Merck
  • Direct catalytic asymmetric addition of allyl cyanide to ketones via soft Lewis acid/hard Brønsted base/hard Lewis base catalysis.
Direct catalytic asymmetric addition of allyl cyanide to ketones via soft Lewis acid/hard Brønsted base/hard Lewis base catalysis.
Journal of the American Chemical Society (2010-03-27)
Ryo Yazaki, Naoya Kumagai, Masakatsu Shibasaki
ABSTRACT
We report that a hard Lewis base substantially affects the reaction efficiency of direct catalytic asymmetric gamma-addition of allyl cyanide (1a) to ketones promoted by a soft Lewis acid/hard Brønsted base catalyst. Mechanistic studies have revealed that Cu/(R,R)-Ph-BPE and Li(OC(6)H(4)-p-OMe) serve as a soft Lewis acid and a hard Brønsted base, respectively, allowing for deprotonative activation of 1a as the rate-determining step. A ternary catalytic system comprising a soft Lewis acid/hard Brønsted base and an additional hard Lewis base, in which the basicity of the hard Brønsted base Li(OC(6)H(4)-p-OMe) was enhanced by phosphine oxide (the hard Lewis base) through a hard-hard interaction, outperformed the previously developed binary soft Lewis acid/hard Brønsted base catalytic system, leading to higher yields and enantioselectivities while using one-tenth the catalyst loading and one-fifth the amount of 1a. This second-generation catalyst allows efficient access to highly enantioenriched tertiary alcohols under nearly ideal atom-economical conditions (0.5-1 mol % catalyst loading and a substrate molar ratio of 1:2).
Sigma-Aldrich
Allyl cyanide, 98%