Merck
  • Effects of diisononyl phthalate on atopic dermatitis in vivo and immunologic responses in vitro.
Effects of diisononyl phthalate on atopic dermatitis in vivo and immunologic responses in vitro.
Environmental health perspectives (2010-01-13)
Eiko Koike, Rie Yanagisawa, Kaori Sadakane, Ken-ichiro Inoue, Takamichi Ichinose, Hirohisa Takano
ABSTRACT
Diisononyl phthalate (DINP), a principal plasticizer in many polyvinyl chloride products, has been shown to have an adjuvant effect on immunoglobulin (Ig) production in mice. However, the effects of DINP on allergic diseases have not been fully elucidated. In the present study we investigated the effects of DINP on atopic dermatitis (AD)-like skin lesions induced by Dermatophagoides pteronyssinus (Dp) in atopic-prone NC/Nga mice. Mice were injected intradermally with Dp on their ears and were exposed to DINP (0, 0.15, 1.5, 15, or 150 mg/kg/day) intraperitoneally. We evaluated clinical scores, ear thickening, histologic findings, protein expression of cytokines/chemokines in the ear, and serum levels of Ig and histamine. Furthermore, we investigated the effects of DINP on bone-marrow-derived dendritic cells (BMDCs) or splenocytes in vitro. After exposure to DINP (0-100 microM), cells were evaluated for phenotype and function. DINP aggravated AD-like skin lesions related to Dp. The aggravation was consistent with eosinophilic inflammation, mast cell degranulation, and thymic stromal lymphopoietin (TSLP) expression in the ear. DINP enhanced the expression of cell surface activation markers on BMDCs and their production of TARC/CCL17 (thymus- and activation-regulated chemokine) and MDC/CCL22 (macrophage-derived chemokine), as well as their capacity to stimulate Dp-specific T-cell proliferation. DINP also enhanced interleukin-4 production and Dp-stimulated proliferation of splenocytes. DINP can aggravate AD-like skin lesions related to Dp. The mechanisms of the aggravation might be mediated, at least partly, through the TSLP-related activation of dendritic cells and by direct or indirect activation of the immune cells.
Sigma-Aldrich
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade