Merck
Microcapsules with three orthogonal reactive sites.
Organic letters (2009-03-04)
Brian P Mason, Steven M Hira, Geoffrey F Strouse, D Tyler McQuade
ABSTRACT
Polymeric microcapsules containing reactive sites on the shell surface and two orthogonally reactive polymers encapsulated within the interior are selectively labeled. The capsules provide three spatially separate and differentially reactive sites. Confocal fluorescence microscopy is used to characterize the distribution of labels. Polymers encapsulated are distributed homogeneously within the core and do not interact with the shell even when oppositely charged.
Sigma-Aldrich
Poly[(phenyl isocyanate)-co-formaldehyde], average Mn ~400
Sigma-Aldrich
Poly[(phenyl isocyanate)-co-formaldehyde], average Mn ~340