Merck
  • Gas chromatography-mass spectrometry assay for determination of Ntau-methylhistamine concentration in canine urine specimens and fecal extracts.
Gas chromatography-mass spectrometry assay for determination of Ntau-methylhistamine concentration in canine urine specimens and fecal extracts.
American journal of veterinary research (2009-02-24)
Craig G Ruaux, Jeannie M Wright, Jörg M Steiner, David A Williams
ABSTRACT
To develop and validate a gas chromatography-mass spectrometry (GC-MS) method for determination of Ntau-methylhistamine (NMH) concentration in canine urine and fecal extracts and to assess urinary NMH concentrations in dogs with mast cell neoplasia and fecal NMH concentrations in dogs with protein-losing enteropathy. Urine specimens were collected from 6 healthy dogs and 7 dogs with mast cell neoplasia. Fecal extracts were obtained from fecal specimens of 28 dogs with various severities of protein-losing enteropathy, as indicated by fecal concentration of alpha1-proteinase inhibitor. NMH was extracted directly from urine, and fecal specimens were first extracted into 5 volumes of PBSS containing 1% newborn calf serum. Ntau-methylhistamine in specimens was quantified via stable isotope dilution GC-MS. The assay was validated via determination of percentage recovery of known amounts of NMH and interassay coefficients of variation. Urinary excretion of NMH was evaluated by means of NMH-to-creatinine concentration ratios. Recovery of NMH in urine and fecal extracts averaged 104.6% and 104.5%, respectively. Interassay coefficients of variation ranged from 5.4% to 11.7% in urine and 12.6% to 18.1% in fecal extracts. Urinary NMH excretion was significantly increased in dogs with mast cell neoplasia, compared with that in healthy dogs. No correlation was detected between severity of protein-losing enteropathy and fecal NMH concentration. This method provided a sensitive, reproducible means of measuring NMH in canine urine and fecal extracts. High urinary NMH-to-creatinine concentration ratios in dogs with mast cell neoplasia are consistent with increased histamine release in this disease.
Product Description
Supelco
3-Methylhistamine dihydrochloride, analytical standard
Pricing and availability is not currently available.