Merck
  • Flow injection analysis of lactose using covalently immobilized beta-galactosidase, mutarotase, and glucose oxidase/peroxidase on a 2-fluoro-1-methylpyridinium salt-activated Fractogel support.
Flow injection analysis of lactose using covalently immobilized beta-galactosidase, mutarotase, and glucose oxidase/peroxidase on a 2-fluoro-1-methylpyridinium salt-activated Fractogel support.
Analytical biochemistry (1991-04-01)
D Narinesingh, V A Stoute, G Davis, T T Ngo
ABSTRACT
Milk samples were analyzed for their lactose content using flow injection analysis and incorporating immobilized beta-galactosidase or beta-galactosidase/mutarotase and glucose oxidase/peroxidase bioreactors. These enzymes were immobilized, under mild conditions, on to a 2-fluoro-1-methylpyridinium salt-activated Fractogel support. The use of a phosphate buffer (0.15 M) was found to facilitate the rapid mutarotation of alpha-D-glucose and hence could obviate the need for the more expensive mutarotase. The chromogenic agents of choice for monitoring the reaction were 3-methyl-2-benzothiazolinone hydrazone and 3-dimethylaminobenzoic acid. Linearity was observed over the concentration range 16-160 micrograms/ml using lactose standards (r = 0.996). Between 30 and 40 milk samples/h can be analyzed. Comparisons are made with existing HPLC and alkaline methylamine methods for a range of milk matrices. The FIA method consistently gives the lowest standard deviations and coefficient of variation for the various milk matrices analyzed.
Sigma-Aldrich
2-Fluoro-1-methylpyridinium p-toluenesulfonate, <5% 2-hydroxy-1-methylpyridinium p-toluenesulfonate, technical grade, ā‰„90%