- First chemical synthesis of antioxidative metabolites of sesamin.
First chemical synthesis of antioxidative metabolites of sesamin.
Chemical & pharmaceutical bulletin (2008-11-05)
Hidehito Urata, Yuka Nishioka, Takafumi Tobashi, Yasuo Matsumura, Namino Tomimori, Yoshiko Ono, Yoshinobu Kiso, Shun-Ichi Wada
PMID18981616
ABSTRACT
The first chemical synthesis of two metabolites ((1R,2S,5R,6S)-6-(3,4-dihydroxyphenyl)-2-(3,4-methylenedioxyphenyl)-3,7-dioxabicyclo[3,3,0]octane (SC-1) and (1R,2S,5R,6S)-2,6-bis(3,4-dihydroxyphenyl)-3,7-dioxabicyclo[3,3,0]octane (SC-2)) of sesamin was achieved by a simple two-step approach from sesamin. The approach consists of acetoxylation of the methylenedioxy moiety(ies) with lead(IV) tetraacetate and acid hydrolysis of the resulting hemiorthoester to SC-1 and SC-2.
MATERIALS