Merck
  • Contribution of alpha4beta1 integrin to the antiallergic effect of levocabastine.
Contribution of alpha4beta1 integrin to the antiallergic effect of levocabastine.
Biochemical pharmacology (2008-08-06)
Ahmed R Qasem, Claudio Bucolo, Monica Baiula, Antonino Spartà, Paolo Govoni, Andrea Bedini, Domenico Fascì, Santi Spampinato
ABSTRACT
Levocabastine is an antiallergic drug acting as a histamine H1-receptor antagonist. In allergic conjunctivitis (AC), it may also antagonize up-regulation of the intercellular adhesion molecule-1 (ICAM-1) expressed on epithelial conjunctival cells. However, little is known about its effects on eosinophils, important effector cells in AC. The adhesion molecule integrin alpha(4)beta(1) is expressed in eosinophils; it interacts with the vascular cell adhesion molecule-1 (VCAM-1) and fibronectin (FN) in vascular endothelial cells and contributes to eosinophil activation and infiltration in AC. This study provides evidence that in a scintillation proximity assay levocabastine (IC(50) 406 microM), but not the first-generation antihistamine chlorpheniramine, displaced (125)I-FN binding to human integrin alpha(4)beta(1) and, in flow cytometry analysis, levocabastine antagonized the binding of a primary antibody to integrin alpha(4) expressed on the Jurkat cell surface. Levocabastine, but not chlorpheniramine, binds the alpha(4)beta(1) integrin and prevents eosinophil adhesion to VCAM-1, FN or human umbilical vascular endothelial cells (HUVEC) in vitro. Similarly, levocabastine affects alpha(L)beta(2)/ICAM-1-mediated adhesion of Jurkat cells. In a model of AC levocabastine eye drops reduced the clinical aspects of the late-phase reaction and the conjunctival expression of alpha(4)beta(1) integrin by reducing infiltrated eosinophils. We propose that blockade of integrin-mediated cell adhesion might be a target of the antiallergic action of levocabastine and may play a role in preventing eosinophil adhesion and infiltration in AC.
Sigma-Aldrich
Levocabastine hydrochloride, ≥99% (HPLC), solid