Merck
  • Di(2-ethylhexyl)phthalate induces hepatic tumorigenesis through a peroxisome proliferator-activated receptor alpha-independent pathway.
Di(2-ethylhexyl)phthalate induces hepatic tumorigenesis through a peroxisome proliferator-activated receptor alpha-independent pathway.
Journal of occupational health (2007-06-19)
Yuki Ito, Osamu Yamanoshita, Nobuyuki Asaeda, Yoshiaki Tagawa, Chul-Ho Lee, Toshifumi Aoyama, Gaku Ichihara, Koichi Furuhashi, Michihiro Kamijima, Frank J Gonzalez, Tamie Nakajima
ABSTRACT
Di(2-ethylhexyl)phthalate (DEHP), a commonly used industrial plasticizer, causes liver tumorigenesis presumably via activation of peroxisome proliferator-activated receptor alpha (PPARalpha). The mechanism of DEHP tumorigenesis has not been fully elucidated, and to clarify whether DEHP tumorigenesis is induced via PPARalpha, we compared DEHP-induced tumorigenesis in wild-type and Pparalpha-null mice. Mice of each genotype were divided into three groups, and treated for 22 months with diets containing 0, 0.01 or 0.05% DEHP. Surprisingly, the incidence of liver tumors was higher in Pparalpha-null mice exposed to 0.05% DEHP (25.8%) than in similarly exposed wild-type mice (10.0%). These results suggest the existence of pathways for DEHP-induced hepatic tumorigenesis that are independent of PPARalpha. The levels of 8-OHdG increased dose-dependently in mice of both genotypes, but the degree of increase was higher in Pparalpha-null than in wild-type mice. NFkappaB levels also significantly increased in a dose-dependent manner in Pparalpha-null mice. The protooncogene c-jun-mRNA was induced, and c-fos-mRNA tended to be induced only in Pparalpha-null mice fed a 0.05% DEHP-containing diet. These results suggest that increases in oxidative stress induced by DEHP exposure may lead to the induction of inflammation and/or the expression of protooncogenes, resulting in a high incidence of tumorigenesis in Pparalpha-null mice.
Sigma-Aldrich
Bis(methylglycol) phthalate, technical, ≥90% (GC)
Supelco
Bis(methylglycol) phthalate, analytical standard