- A new class of nifuroxazide analogues: synthesis of 5-nitrothiophene derivatives with antimicrobial activity against multidrug-resistant Staphylococcus aureus.
A new class of nifuroxazide analogues: synthesis of 5-nitrothiophene derivatives with antimicrobial activity against multidrug-resistant Staphylococcus aureus.
Bioorganic & medicinal chemistry (2007-04-10)
Andrea Masunari, Leoberto Costa Tavares
PMID17419064
ABSTRACT
Hospital-acquired methicillin-resistant Staphylococcus aureus (MRSA) has been an increasing problem worldwide since the initial reports over 40 years ago. To examine new drug leads with potential antibacterial activities, 14 p-substituted benzoic acid [(5-nitro-thiophen-2-yl)-methylene]-hydrazides were designed, synthesized, and tested against standard and multidrug-resistant S. aureus strains by serial dilution tests. All compounds exhibited significant bacteriostatic activity and some of them also showed bactericidal activity. The results confirmed the potential of this class of compounds as an alternative for the development of selective antimicrobial agents.