Merck
  • Column switching liquid chromatography and post-column photochemically fluorescence detection to determine imidacloprid and 6-chloronicotinic acid in honeybees.
Column switching liquid chromatography and post-column photochemically fluorescence detection to determine imidacloprid and 6-chloronicotinic acid in honeybees.
Journal of chromatography. A (2007-02-28)
M D Gil García, M Martínez Galera, R Santiago Valverde, A Galanti, S Girotti
ABSTRACT
The determination of imidacloprid and its main metabolite (6-chloronicotinic acid) in honeybees was performed by liquid chromatography with post-column photochemical derivatisation in alkaline medium and fluorescence detection. The compounds were extracted from honeybees with acetone under ultrasound conditions prior to liquid-liquid partition with dichloromethane. The separation of extract components was performed using a 50 mm x 4.6 mm i.d. short column packed with 5 microm Aquasil C(18) using an acetonitrile:water gradient program as mobile phase. Injection of samples in 0.1 mol L(-1) H(3)PO(4)/KH(2)PO(4) buffer solution (pH 3) improved the chromatographic separation between the most polar components of matrix and the 6-chloronicotinic acid. Matrix components were removed to waste using an on-line clean-up method previously to post-column reaction. Limits of quantification were 0.3 and 5.0 microg L(-1) (corresponding to 1.2 and 20.0 microg kg(-1) in the honeybee sample) for imidacloprid and 6-chloronicotinic acid, respectively. The recovery was ranged from 80.2 to 91.7% with a relative standard deviation lower than 9.0%.
Product Description
Supelco
6-Chloropyridine-3-carboxylic acid, PESTANAL®, analytical standard
Sigma-Aldrich
6-Chloropyridine-3-carboxylic acid, 99%