Merck
Coiled molecules in spring loaded devices.
Journal of the American Chemical Society (2006-11-23)
Dariush Ajami, Julius Rebek
ABSTRACT
A cylindrical capsule containing a coiled alkane incorporates spacers. The alkane lengthens and gauche strain is relieved. The spacer can be removed to reset the system to the original state. The system represents a spring-loaded device that operates reversibly in response to acids and bases.
Sigma-Aldrich
Glycoluril, 97%