Merck
  • Homogeneous, competitive fluorescence quenching immunoassay based on gold nanoparticle/polyelectrolyte coated latex particles.
Homogeneous, competitive fluorescence quenching immunoassay based on gold nanoparticle/polyelectrolyte coated latex particles.
The journal of physical chemistry. B (2006-07-21)
Noritaka Kato, Frank Caruso
ABSTRACT
This study reports a homogeneous and competitive fluorescence quenching immunoassay based on gold nanoparticle/polyelectrolyte (Au(NP)/PE) coated latex particles prepared by the layer-by-layer (LbL) technique. First, the resonant energy transfer from a layer of fluorescent PEs to Au(NP) in LbL assembled films on planar substrates was investigated. The quenching efficiency (QE) for the planar films depended on the cube of the distance between the two layers. A QE of 50% was achieved at a distance of ca. 15 nm, indicating that the Au(NP)/PE system is suitable for detecting binding/release events for antibodies. A homogeneous, competitive binding immunoassay for biotin was designed based on Au(NP)/PE-coated polystyrene particles of 488 nm diameter as quenching agents for a fluorescein isothiocyanate labeled anti-biotin immunoglobulin (FITC-anti-biotin IgG). Biotin molecules were localized on the Au(NP)/PE-coated latexes by depositing a layer of biotinylated poly(allylamine hydrochloride) (B-PAH), and FITC-anti-biotin IgGs were subsequently bound to the particles through interaction with the biotin on B-PAH. Transmission electron microscopy and quartz crystal microgravimetry confirmed the multilayer formation on latex particles and planar gold surfaces, respectively. The biotin-functionalized Au(NP)/PE-coated latexes terminated by FITC-anti-biotin IgG exhibited a dynamic sensing range of 1-50 nmol. These results indicate that Au(NP)/PE-coated latexes can be readily used as dynamic range tunable sensors.
Sigma-Aldrich
2-(4-Hydroxyphenylazo)benzoic acid, ≥90% (HPLC)
Supelco
2-(4-Hydroxyphenylazo)benzoic acid, matrix substance for MALDI-MS, ≥99.5%