- Incidence and pathogenic effect of Streptococcus pseudopneumoniae.
Incidence and pathogenic effect of Streptococcus pseudopneumoniae.
Journal of clinical microbiology (2006-06-08)
Colette Harf-Monteil, Carole Granello, Cécile Le Brun, Henri Monteil, Philippe Riegel
PMID16757628
ABSTRACT
We evaluated the incidence of Streptococcus pseudopneumoniae in clinical isolates by phenotypic methods and DNA-DNA hybridization. The pathogenic role of this organism was investigated with the mouse peritonitis/sepsis model. Our results show a low incidence (1/120 pneumococcal isolates) and a potential pathogenic effect for S. pseudopneumoniae.