Merck
  • Effect of solvent on the lithium-bromine exchange of aryl bromides: reactions of n-butyllithium and tert-butyllithium with 1-bromo-4-tert-butylbenzene at 0 degrees C.
Effect of solvent on the lithium-bromine exchange of aryl bromides: reactions of n-butyllithium and tert-butyllithium with 1-bromo-4-tert-butylbenzene at 0 degrees C.
The Journal of organic chemistry (2006-03-25)
William F Bailey, Mark R Luderer, Kevin P Jordan
ABSTRACT
The outcome of reactions of 1-bromo-4-tert-butylbenzene (1), a representative aryl bromide, with n-BuLi or t-BuLi at 0 degrees C in a variety of solvent systems has been investigated. The products of reactions of 1 with n-BuLi vary significantly with changes in solvent composition: 1 does not react with n-BuLi in pure heptane; the exchange reaction to give (4-tert-butylphenyl)lithium, which is slow in pure diethyl ether, is virtually quantitative in heptane containing a small quantity of THF; and the reaction of 1 with n-BuLi in THF leads to considerable coupling. Lithium-bromine exchange is the virtually exclusive outcome of reactions of 1 with t-BuLi in every solvent studied except pure heptane: the presence of a small quantity of any of a variety of structurally diverse ethers (Et(2)O, THF, THP, MTBE) in the predominantly hydrocarbon medium affords (4-tert-butylphenyl)lithium, assayed as tert-butylbenzene, in yields exceeding 97%. The only side products observed from reactions of 1 with t-BuLi are small amounts of benzyne-derived hydrocarbons.
Sigma-Aldrich
tert-Butylbenzene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Bromo-4-tert-butylbenzene, 97%
Pricing and availability is not currently available.