Merck
  • N-triazinylammonium tetrafluoroborates. A new generation of efficient coupling reagents useful for peptide synthesis.
N-triazinylammonium tetrafluoroborates. A new generation of efficient coupling reagents useful for peptide synthesis.
Journal of the American Chemical Society (2005-12-01)
Zbigniew J Kamiński, Beata Kolesińska, Justyna Kolesińska, Giuseppina Sabatino, Mario Chelli, Paolo Rovero, Michał Błaszczyk, Marek L Główka, Anna Maria Papini
ABSTRACT
A new generation of triazine-based coupling reagents (TBCRs), designed according to the concept of "superactive esters", was obtained by treatment of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium (DMTMM) chloride with lithium or silver tetrafluoroborate. The structure of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate was confirmed by X-ray diffraction. Activation of carboxylic acids by using this reagent proceeds via triazine "superactive ester". The coupling reagent was successfully used for the synthesis of Z-, Boc-, and Fmoc-protected dipeptides derived from natural and unnatural sterically hindered amino acids and for fragment condensation, in 80-100% yield and with high enantiomeric purity. The manual SPPS of the ACP(65-74) peptide fragment (H-Val-Gln-Ala-Ala-Ile-Asp-Tyr-Ile-Asn-Gly-OH) proceeded significantly faster than with TBTU or HATU, as well as the automated SPPS of the same fragment gave a purer product than by using TBTU or PyBOP. The reagent was also demonstrated to be efficient in on-resin head-to-tail cyclization of constrained cyclopeptides, in SPPS synthesis of Aib peptides, and in the synthesis of esters from appropriate acids, alcohols, and phenols. The high efficiency and versatility of this new generation of TBCRs confirm, for the first time, the usefulness of the concept of "superactive esters" in rational design of the structure of coupling reagents.
Sigma-Aldrich
Silver tetrafluoroborate, ≥99.99% trace metals basis
Sigma-Aldrich
Silver tetrafluoroborate, 98%