Merck
  • Effect of oral nitroethane and 2-nitropropanol administration on methane-producing activity and volatile fatty acid production in the ovine rumen.
Effect of oral nitroethane and 2-nitropropanol administration on methane-producing activity and volatile fatty acid production in the ovine rumen.
Bioresource technology (2005-11-24)
R C Anderson, G E Carstens, R K Miller, T R Callaway, C L Schultz, T S Edrington, R B Harvey, D J Nisbet
ABSTRACT
Strategies are sought to reduce economic and environmental costs associated with ruminant methane emissions. The effect of oral nitroethane or 2-nitropropanol administration on ruminal methane-producing activity and volatile fatty acid production was evaluated in mature ewes. Daily administration of 24 and 72 mg nitroethane/kg body weight reduced (P<0.05) methane-producing activity by as much as 45% and 69% respectively, when compared to control animals given no nitroethane. A daily dose of 120 mg 2-nitropropanol/kg body weight was needed to reduce (P<0.05) methane-producing activity by 37% from that of untreated control animals. Reductions in methane-producing activity may have been diminished by the last day (day 5) of treatment, presumably due to ruminal adaptation. Oral administration of nitroethane or 2-nitropropanol had little or no effect on accumulations or molar proportions of volatile fatty acids in ruminal contents collected from the sheep. These results demonstrate that nitroethane was superior to 2-nitropropanol as a methane inhibitor and that both nitrocompounds reduced ruminal methanogenesis in vivo without redirecting the flow of reductant generated during fermentation to propionate and butyrate.
Sigma-Aldrich
Nitroethane, reagent grade, ≥98.0%
Sigma-Aldrich
Nitroethane, ReagentPlus®, 99.5%