- Endoplasmic reticulum retention of the large splice variant of the UDP-galactose transporter is caused by a dilysine motif.
Endoplasmic reticulum retention of the large splice variant of the UDP-galactose transporter is caused by a dilysine motif.
Nucleotide-sugar transporters supply mainly the Golgi glycosyltransferases with substrates. Some glycosyltransferases in the endoplasmic reticulum (ER), however, also use activated sugars. Recent studies have demonstrated that UDP-galactose (UDP-Gal) is the substrate for the ER resident ceramide-galactosyltransferase (cer-GalT) and cells expressing cer-GalT are able to retain the UDP-Gal transporter (UGT) by physical contacts formed between the two proteins. Here, we describe a second active mechanism for ER localization of the UGT. The UGT is produced in two splice forms UGT1 and UGT2. The proteins vary only at their extreme C-termini but show strikingly different intracellular distribution. Although N-terminally epitope tagged forms of UGT1 localize exclusively to the Golgi, similar constructs of UGT2 show both ER and Golgi localization. The dilysine motif KVKGS contained in UGT2 can be demonstrated to be responsible for the dual localization because: (1) disturbance of the signal via site specific mutation or C-terminal extension completely shifts the transporter to the Golgi, (2) transfer of the dilysine motif is sufficient to redistribute the Golgi CMP-sialic acid transporter to the ER, and (3) replacement of KVKGS by the strong ER retention signal KKNT is sufficient to completely retain UGT2 in the ER.