Merck
  • Multiresidue determination of pesticides in malt beverages by capillary gas chromatography with mass spectrometry and selected ion monitoring.
Multiresidue determination of pesticides in malt beverages by capillary gas chromatography with mass spectrometry and selected ion monitoring.
Journal of agricultural and food chemistry (2004-10-14)
Jon W Wong, Michael G Webster, Dawit Z Bezabeh, Mathew J Hengel, Kenley K Ngim, Alexander J Krynitsky, Susan E Ebeler
ABSTRACT
A method was developed to determine pesticides in malt beverages using solid phase extraction on a polymeric cartridge and sample cleanup with a MgSO4-topped aminopropyl cartridge, followed by capillary gas chromatography with electron impact mass spectrometry in the selected ion monitoring mode [GC-MS(SIM)]. Three GC injections were required to analyze and identify organophosphate, organohalogen, and organonitrogen pesticides. The pesticides were identified by the retention times of peaks of the target ion and qualifier-to-target ion ratios. GC detection limits for most of the pesticides were 5-10 ng/mL, and linearity was determined from 50 to 5000 ng/mL. Fortification studies were performed at 10 ng/mL for three malt beverages that differ in properties such as alcohol content, solids, and appearance. The recoveries from the three malt beverages were greater than 70% for 85 of the 142 pesticides (including isomers) studied. The data showed that the different malt beverage matrixes had no significant effect on the recoveries. This method was then applied to the screening and analysis of malt beverages for pesticides, resulting in the detection of the insectide carbaryl and the fungicide dimethomorph in real samples. The study indicates that pesticide levels in malt beverages are significantly lower than the tolerance levels set by the United States Environmental Protection Agency for malt beverage starting ingredients. The use of the extraction/cleanup procedure and analysis by GC-MS(SIM) proved effective in screening malt beverages for a wide variety of pesticides.
Product Description
Supelco
Dimethomorph, PESTANAL®, analytical standard