Merck
  • Synthesis of an apiose-containing disaccharide fragment of rhamnogalacturonan-II and some analogues.
Synthesis of an apiose-containing disaccharide fragment of rhamnogalacturonan-II and some analogues.
Carbohydrate research (2003-12-09)
Anne-Laure Chauvin, Sergey A Nepogodiev, Robert A Field
ABSTRACT
Beta-rhamnosylation of methyl 2-C-hydroxymethyl-2,3-O-isopropylidene-beta-D-erythrofuranoside and methyl 2,3-O-isopropylidene-beta-D-ribofuranoside was achieved using 4-O-acetyl-2,3-O-carbonyl-alpha-L-rhamnopyranosyl bromide and Ag2O as a promoter. Deprotected disaccharides beta-L-Rhap-(1-->3')-beta-D-Apif-OMe and beta-L-Rhap-(1-->3')-beta-D-Ribf-OMe were compared to their alpha-rhamnosyl isomers which were prepared using conventional Helferich glycosylation.
Sigma-Aldrich
D-Apiose solution, ≥99.0% (HPLC)