- [Antidotal effects of 2,3-dimercaptopropane-1-sulfonate sodium (DMPS) and combined with diazepam on acute poisoning caused by sodium ammonium dimethyl-2-propano-1,3-dithiosulfate monohydrate (SCD)].
[Antidotal effects of 2,3-dimercaptopropane-1-sulfonate sodium (DMPS) and combined with diazepam on acute poisoning caused by sodium ammonium dimethyl-2-propano-1,3-dithiosulfate monohydrate (SCD)].
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine] (1992-07-01)
Z Q Lu, G X Hu, Z K Chen
PMID1338778
ABSTRACT
In mice, DMPS (250 mg/kg, i.v.) combined with diazepam (1.25 mg/kg, i.p.) could increase LD50 of p. o. SCD 5.3 times. DMPS (62.5 mg/kg, i.v.) antagonized completely the respiratory depression and neuromuscular blockade caused by SCD(7.5 mg/kg, i.v.) in rabbits. SCD (15 mg/kg, i.v.) caused tremor, tonic convulsion and the abnormal paroxysmal discharges in EEG in rabbits. DMPS (0.5 mg/kg, i.c.v) could not eliminate the abnormal paroxysmal discharges in EEG of rabbits. DMPS (62.5 mg/kg, i.v.) combined with diazepam (5 mg/kg, i.v.) completely and rapidly antagonize these toxic symptoms and the abnormal changes in EEG.