Merck
  • Characteristic odor of Osmoderma eremita identified as a male-released pheromone.
Characteristic odor of Osmoderma eremita identified as a male-released pheromone.
Journal of chemical ecology (2003-05-22)
Mattias C Larsson, Jonas Hedin, Glenn P Svensson, Till Tolasch, Wittko Francke
ABSTRACT
Osmoderma eremita (Scopoli) is an endangered scarab beetle living in hollow trees. It has mainly been known for its characteristic odor, typically described as a fruity, peachlike or plumlike aroma. The odor emanating from a single beetle can sometimes be perceived from a distance of several meters. In this paper, we show that the characteristic odor from O. eremita is caused by the compound (R)-(+)-gamma-decalactone, released in large quantities mainly or exclusively by male beetles. Antennae from male and female beetles responded in a similar way to (R)-(+)-gamma-decalactone in electroantennographic recordings. Field trapping experiments showed that (R)-(+)-gamma-decalactone is a pheromone attracting female beetles. Lactones similar to (R)-(+)-gamma-decalactone are frequently used as female-released sex pheromones by phytophagous scarabs. This is, however, the first evidence of a lactone used as a male-produced pheromone in scarab beetles. We propose that the strong signal from males is a sexually selected trait used to compete for females and matings. The signal could work within trees but also act as a guide to tree hollows, which are an essential resource for O. eremita. Males may, thus, attract females dispersing from their natal tree by advertising a suitable habitat. This signal could also be exploited by other males searching for tree hollows or for females, which would explain the catch of several males in our traps.
Sigma-Aldrich
γ-Decalactone, ≥98%, FCC, FG
Sigma-Aldrich
γ-Decanolactone, 98%