- [Advances in the detection method of several forbidden rodenticides].
[Advances in the detection method of several forbidden rodenticides].
Wei sheng yan jiu = Journal of hygiene research (2003-02-26)
Weiping Xie, Zhonghui Chen, Xiaohua Li
PMID12600049
ABSTRACT
The progress of detection method of four forbidden rodenticides, including fluoroacetamide, sodium fluoroacetate, gliftor and tetramine is reviewed in this paper. The technique of sample preparation and gas chromatography are emphasized.