Merck
Bioavailability and biodegradation of prosulfocarb in soil.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2002-06-26)
Mara Gennari, Roberto Ambrosoli, Michèle Nègre, José Luis Minati
ABSTRACT
Active microbial degraders of the herbicide prosulfocarb (PSC) were isolated to evaluate their performance in soil with a view to their use for bioremediation. The isolated cultures (a microbial consortium and a Pseudomonas sp. strain) were active when tested in mineral medium with PSC as the only carbon source, but had an adverse effect on the soil indigenous microflora. Biodegradation in the inoculated soils was thus lower than in the uninoculated soil when only the indigenous microflora was present. Further tests showed that the strong affinity of PSC for soil organic matter affected its bioavailability and hence its biodegradation by the inocula. Bioremediation of PSC contaminated soils could thus be undertaken by biostimulation of indigenous microflora.
Product Description
Supelco
Prosulfocarb, PESTANAL®, analytical standard
Pricing and availability is not currently available.