- Use of thallium(I) ethoxide in Suzuki cross coupling reactions.
Use of thallium(I) ethoxide in Suzuki cross coupling reactions.
Organic letters (2000-09-16)
S A Frank, H Chen, R K Kunz, M J Schnaderbeck, W R Roush
PMID10990429
ABSTRACT
[reaction: see text]Thallium(I) ethoxide promotes Suzuki cross couplings for a range of vinyl- and arylboronic acids with vinyl and aryl halide partners in good to excellent yields. This reagent offers distinct advantages over thallium(I) hydroxide in terms of commercial availability, stability, and ease of use.
MATERIALS