Merck
Biologically stable [(18)F]-labeled benzylfluoride derivatives.
Nuclear medicine and biology (2000-04-25)
Y Magata, L Lang, D O Kiesewetter, E M Jagoda, M A Channing, W C Eckelman
ABSTRACT
Use of the [(18)F]-fluoromethyl phenyl group is an attractive alternative to direct fluorination of phenyl groups because the fluorination of the methyl group takes place under milder reaction conditions. However, we have found that 4-FMeBWAY showed femur uptake equal to that of fluoride up to 30 min in rat whereas 4-FMeQNB had a significantly lower percent injected dose per gram in femur up to 120 min. For these and other benzylfluoride derivatives, there was no clear in vivo structure-defluorination relationship. Because benzylchlorides (BzCls) are known alkylating agents, benzylfluorides may be alkylating agents as well, which may be the mechanism of defluorination. On this basis, the effects of substitution on chemical stability were evaluated by the 4-(4-nitro-benzyl)-pyridine (NBP) test, which is used to estimate alkylating activity with NBP. The effect of substitution on the alkylating activity was evaluated for nine BzCl derivatives: BzCl; 3- or 4-methoxy (electron donation) substituted BzCl; 2-, 3-, or 4-nitro (electron withdrawing) substituted BzCl; and 2-, 3-, or 4-chloro (electron withdrawing) substituted BzCl. Taken together, the alkylating reactivity of 3-chloro-BzCl was the weakest. This result was then applied to [(18)F]-benzylfluoride derivatives and in vivo and in vitro stability were evaluated. Consequently, 3-chloro-[(18)F]-benzylfluoride showed a 70-80% decrease of defluorination in both experiments in comparison with [(18)F]-benzylfluoride, as expected. Moreover, a good linear relationship between in vivo femur uptake and in vitro hepatocyte metabolism was observed with seven (18)F-labeled radiopharmaceuticals, which were benzylfluorides, alkylfluorides, and arylfluorides. Apparently, the [(18)F]-fluoride ion is released by metabolism in the liver in vivo. In conclusion, 3-chloro substituted BzCls are the most stable, which suggests that 3-chloro benzylfluorides will be the most chemically stable compound. This result should be important in future design of radioligands labeled with a benzylfluoride moiety.
Sigma-Aldrich
3-Bromobenzyl chloride, 98%
Sigma-Aldrich
4-(4-Nitrobenzyl)pyridine, 98%