Merck
  • Immobilization of tripeptide growth factor glycyl-L-histidyl-L-lysine on poly(vinylalcohol)-quarternized stilbazole (PVA-SbQ) and its use as a ligand for hepatocyte attachment.
Immobilization of tripeptide growth factor glycyl-L-histidyl-L-lysine on poly(vinylalcohol)-quarternized stilbazole (PVA-SbQ) and its use as a ligand for hepatocyte attachment.
Biological & pharmaceutical bulletin (1999-10-08)
M Kawase, N Miura, N Kurikawa, K Masuda, S Higashiyama, K Yagi, T Mizoguchi
ABSTRACT
A tripeptide growth factor, glycyl-L-histidyl-L-lysine (GHK), was immobilized on the surface of poly(vinylalcohol)-quarternized stilbazole (PVA-SbQ) gel. The photoreactive substance, 4-(3-trifluoromethylazirino)benzoyl-N-hydroxysuccinimide (TDBA-OSu), was employed to link the gel and ligand GHK. The density of immobilized GHK was 70 nmol/cm2. Isolated rat hepatocytes were inoculated on the GHK-immobilized PVA-SbQ gel and cultured for 5 d. About 24 h after inoculation, hepatocytes started to aggregate and formed multicellular spheroids while almost no cells attached to GHK-non-immobilized PVA-SbQ gel. The formed spheroids attached firmly to the surface of PVA-SbQ gel for 5 d. GHK was, thus, shown to be an effective ligand for hepatocyte attachment. Dodecamethylenediamine was used to extend the length between the gel surface and GHK. Extension of the length significantly increased the number of attached hepatocytes.
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)
Pricing and availability is not currently available.