- Inhibition of ICE family proteases by baculovirus antiapoptotic protein p35.
Inhibition of ICE family proteases by baculovirus antiapoptotic protein p35.
The baculovirus antiapoptotic protein p35 inhibited the proteolytic activity of human interleukin-1 beta converting enzyme (ICE) and three of its homologs in enzymatic assays. Coexpression of p35 prevented the autoproteolytic activation of ICE from its precursor form and blocked ICE-induced apoptosis. Inhibition of enzymatic activity correlated with the cleavage of p35 and the formation of a stable ICE-p35 complex. The ability of p35 to block apoptosis in different pathways and in distantly related organisms suggests a central and conserved role for ICE-like proteases in the induction of apoptosis.