Merck
  • DFF45/ICAD can be directly processed by granzyme B during the induction of apoptosis.
DFF45/ICAD can be directly processed by granzyme B during the induction of apoptosis.
Immunity (2000-07-14)
D A Thomas, C Du, M Xu, X Wang, T J Ley
ABSTRACT
Granzyme B (GzmB) is a component of cytotoxic lymphocyte granules that can rapidly initiate apoptosis in target cells. While several procaspases are cleaved and activated by GzmB, the absolute requirement of caspase activation for GzmB-induced apoptosis is controversial. In this report, we demonstrate that GzmB can initiate apoptosis in the absence of caspase-3 activity by directly cleaving DFF45/ICAD to liberate activated DFF40/CAD. DFF45/ICAD cleavage occurs less efficiently in cells that lack caspase-3 activity, suggesting that the caspases normally amplify the GzmB death signal. DFF45/ICAD-deficient mouse embryo fibroblasts are partially resistant to GzmB-induced death, demonstrating the biological importance of DFF45/ICAD for GzmB-mediated apoptosis.
Sigma-Aldrich
Granzyme B from mouse, ≥90% (SDS-PAGE), recombinant, expressed in Pichia pastoris, buffered aqueous solution, ≥10,000 units/mg protein