Merck
  • Isoenzyme specificity of bisindolylmaleimides, selective inhibitors of protein kinase C.
Isoenzyme specificity of bisindolylmaleimides, selective inhibitors of protein kinase C.
The Biochemical journal (1993-09-01)
S E Wilkinson, P J Parker, J S Nixon
ABSTRACT
The protein kinase C (PKC) family of isoenzymes is believed to mediate a wide range of signal-transduction pathways in many different cell types. A series of bisindolylmaleimides have been evaluated as inhibitors of members of the conventional PKC family (PKCs-alpha, -beta, -gamma) and of a representative of the new, Ca(2+)-independent, PKC family, PKC-epsilon. In contrast with the indolocarbazole staurosporine, all the bisindolylmaleimides investigated showed slight selectivity for PKC-alpha over the other isoenzymes examined. In addition, bisindolylmaleimides bearing a conformationally restricted side-chain were less active as inhibitors of PKC-epsilon. Most noticeable of these was Ro 32-0432, which showed a 10-fold selectivity for PKC-alpha and a 4-fold selectivity for PKC-beta I over PKC-epsilon.
Sigma-Aldrich
Ro 31-0432, solid