Merck
  • [Impact of xenoimplants on metabolic changes in the liver of rats in combined trauma].
[Impact of xenoimplants on metabolic changes in the liver of rats in combined trauma].
Klinichna khirurhiia (2013-05-31)
S R Pidruchna
ABSTRACT
In recent years there are intensively studied the role of hepatocytic affection in the complications progress in trauma of various genesis, including the multiple and combined. Positive correcting impact of lyophilized xenoimplants on the hepatic markers activity was demonstrated in the animal blood plasm while severe and combined trauma. Activity of alkaline phosphatase and gammaglutamyltranspeptidase in laboratory animals in polytrauma have had reduced essentially after correction performance during 1 week, and while combined trauma--on the 3rd day.