Merck
  • Adenosine deaminase activity and immunoglobulin levels as potential systemic biomonitors of occupational hazards and health status in municipal solid waste management workers.
Adenosine deaminase activity and immunoglobulin levels as potential systemic biomonitors of occupational hazards and health status in municipal solid waste management workers.
Environmental toxicology and pharmacology (2012-12-12)
Adesina O Odewabi, Omobola A Ogundahunsi, Adenike A Odewabi, Kolawole S Oritogun, Martins Ekor
ABSTRACT
Immune status of waste management workers (WMW) with underlying systemic inflammation was assessed to identify useful immune-related biomarkers of occupational health and safety. Clinical history of WMW revealed high prevalence of respiratory symptoms alongside gastrointestinal and musculoskeletal complaints relative to control. Systemic inflammation, characterized by significant (p<0.001) elevation of erythrocyte sedimentation rate and C-reactive protein, was associated with marked increase in concentration and prevalence of IgA (p<0.05), IgG (p<0.01) and adenosine deaminase activity (ADA) (p<0.01) in WMW. Haematological changes include significant (p<0.01) increase in lymphocytes, monocytes and total leukocytes. Eosinophils also increased significantly (p<0.001) while haemoglobin, packed cell volume and neutrophil decreased significantly (p<0.05). Receiver operating characteristic curve and multivariate analyses revealed ADA (p<0.002) and IgG (p<0.05) as important immune markers respectively for assessing sub-clinical effects of occupational exposure. Our data suggest ADA and IgG as useful immune health and safety indicators in WMW.
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein
Sigma-Aldrich
Adenosine deaminase human, recombinant, expressed in E. coli, ≥1 U/mL