Merck
  • Cloning, heterologous expression, and characterization of novel protease-resistant α-galactosidase from new Sphingomonas strain.
Cloning, heterologous expression, and characterization of novel protease-resistant α-galactosidase from new Sphingomonas strain.
Journal of microbiology and biotechnology (2012-11-06)
Junpei Zhou, Yanyan Dong, Junjun Li, Rui Zhang, Xianghua Tang, Yuelin Mu, Bo Xu, Qian Wu, Zunxi Huang
ABSTRACT
The α-galactosidase-coding gene agaAJB13 was cloned from Sphingomonas sp. JB13 showing 16S rDNA (1,343 bp) identities of < or =97.2% with other identified Sphingomonas strains. agaAJB13 (2,217 bp; 64.9% GC content) encodes a 738-residue polypeptide (AgaAJB13) with a calculated mass of 82.3 kDa. AgaAJB13 showed the highest identity of 61.4% with the putative glycosyl hydrolase family 36 α-galactosidase from Granulicella mallensis MP5ACTX8 (EFI56085). AgaAJB13 also showed <37% identities with reported protease-resistant or Sphingomonas alpha-galactosidases. A sequence analysis revealed different catalytic motifs between reported Sphingomonas α-galactosidases (KXD and RXXXD) and AgaAJB13 (KWD and SDXXDXXXR). Recombinant AgaAJB13 (rAgaAJB13) was expressed in Escherichia coli BL21 (DE3). The purified rAgaAJB13 was characterized using p-nitrophenyl-α-D-galactopyranoside as the substrate and showed an apparent optimum at pH 5.0 and 60 degrees C and strong resistance to trypsin and proteinase K digestion. Compared with reported proteaseresistant alpha-galactosidases showing thermolability at 50 degrees C or 60°C and specific activities of <71 U/mg with or without protease treatments, rAgaAJB13 exhibited a better thermal stability (half-life of >60 min at 60°C) and higher specific activities (225.0-256.5 U/mg). These sequence and enzymatic properties suggest AgaAJB13 is the first identified and characterized Sphingomonas α-galactosidase, and shows novel protease resistance with a potential value for basic research and industrial applications.
Sigma-Aldrich
α-Galactosidase from green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein
Sigma-Aldrich
α-Galactosidase I, Alkaline from Cucumis melo
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution