Merck
  • [Differential expression of haemolymph proteome of resistant strain and susceptible strain for BmNPV in Bombyx mori L].
[Differential expression of haemolymph proteome of resistant strain and susceptible strain for BmNPV in Bombyx mori L].
Sheng wu gong cheng xue bao = Chinese journal of biotechnology (2008-05-10)
Keya Cai, Keping Chen, Xiaoyong Liu, Qin Yao, Jun Li
ABSTRACT
Three model silkworms, highly resistant strain, highly susceptible strain and their near isogenic line were established by hybridization and backcross. The resistance of silkworm (Bombyx mori L.) to BmNPV was studied at proteomic level using two-dimensional gel electrophoresis and MALDI TOF/TOF MS. Differential expression profiles of proteome in resistant strain, susceptible strain and near isogenic line were obtained, and 180, 190 and 187 of protein spots were shown, respectively. Among them, 80% were concentrated in pI 5-9. Twelve differential protein spots in total were obtained from 3 gels. Using MALDI TOF/TOF MS, five proteins, including aminoacylase, were identified from these spots. The exclusive expression of aminoacylase in highly resistant strain and near isogenic line and its absence in susceptible strain suggest that it might be a BmNPV-resistance related protein.
Sigma-Aldrich
Acylase I from Aspergillus melleus, powder, brown, >0.5 U/mg
Sigma-Aldrich
Acylase I, immobilized on Eupergit® C from Aspergillus sp., ≥50 U/g moist material
Sigma-Aldrich
Acylase I from porcine kidney, Grade II, salt-free, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
Acylase I from porcine kidney, Grade I, lyophilized powder, ≥1500 units/mg protein