Merck
  • Saturation mutagenesis of Asn152 reveals a substrate selectivity switch in P99 cephalosporinase.
Saturation mutagenesis of Asn152 reveals a substrate selectivity switch in P99 cephalosporinase.
Protein science : a publication of the Protein Society (2007-11-22)
Scott T Lefurgy, René M de Jong, Virginia W Cornish
ABSTRACT
In class C beta-lactamases, the strictly conserved Asn152 forms part of an extended active-site hydrogen-bonding network. To probe its role in catalysis, all 19 mutants of Enterobacter cloacae P99 cephalosporinase Asn152 were simultaneously constructed and screened in Escherichia coli for their in vivo activity. The screen identified the previously uncharacterized mutants Asn152Ser, Asn152Thr, and Asn152Gly, which possess significant activity and altered substrate selectivity. In vitro measurement of Michaelis-Menten kinetic constants revealed that the Asn152Ser mutation causes a selectivity switch for penicillin G versus cefoxitin. Asn152Thr showed a 63-fold increase in k (cat) for oxacillin, a slow substrate for wild-type cephalosporinase. The results contribute to a growing body of data showing that mutation of highly conserved residues in the active site can result in substrate selectivity changes. The library screening method presented here would be applicable to substrate selectivity determination in other readily screenable enzymes.
Sigma-Aldrich
JumpStart REDAccuTaq® LA DNA Polymerase, Long and accurate hot-start Taq with inert dye, 10X buffer included
Pricing and availability is not currently available.