Merck
  • Kinetics of the M-intermediate in the photocycle of bacteriorhodopsin upon chemical modification with surfactants.
Kinetics of the M-intermediate in the photocycle of bacteriorhodopsin upon chemical modification with surfactants.
Photochemistry and photobiology (2009-12-17)
Li-Kang Chu, Mostafa A El-Sayed
ABSTRACT
The spectroscopic and kinetic studies of the interaction between bacteriorhodopsin in the M-intermediate and several surfactants (cetyl trimethyl ammonium bromide, dodecyl trimethyl ammonium bromide, diethylene glycol mono-n-hexyl ether, ethylene glycol mono-n-hexyl ether, sodium 1-decanesulfonate and sodium 1-heptanesulfonate) have been investigated using steady-state UV-VIS spectrometry and time-resolved absorption techniques. The steady-state spectral results show that bR retains its trimeric state. Time-resolved observations indicate that the rate of deprotonation of the protonated Schiff base increases in the presence of the cationic surfactants, whereas insignificant changes are observed in the neutral or anionic surfactants. The rate of the reprotonation of the Schiff base in the transition M --> N is accelerated in anionic and neutral surfactants, but is decelerated in the presence of the cationic surfactants. Surfactants with a longer hydrocarbon tail have a greater effect on the kinetics when compared with surfactants having shorter hydrocarbon tails. The opposite effect is observed when the hydrophilic head of the surfactants contains opposite charges. These distinct kinetics are discussed in terms of the difference in the modified surface hydrophilicity of the bR and the possible protein configurational changes upon surfactant treatments.
Sigma-Aldrich
Sodium 1-decanesulfonate, BioXtra, ~98%