Merck
  • Oval cell response is attenuated by depletion of liver resident macrophages in the 2-AAF/partial hepatectomy rat.
Oval cell response is attenuated by depletion of liver resident macrophages in the 2-AAF/partial hepatectomy rat.
PloS one (2012-04-20)
Shuai Xiang, Han-Hua Dong, Hui-Fang Liang, Song-Qing He, Wei Zhang, Chang-Hai Li, Bi-Xiang Zhang, Bin-Hao Zhang, Kai Jing, Stephen Tomlinson, Nico van Rooijen, Li Jiang, Katherine Cianflone, Xiao-Ping Chen
ABSTRACT
Macrophages are known to play an important role in hepatocyte mediated liver regeneration by secreting inflammatory mediators. However, there is little information available on the role of resident macrophages in oval cell mediated liver regeneration. In the present study we aimed to investigate the role of macrophages in oval cell expansion induced by 2-acetylaminofluorene/partial hepatectomy (2-AAF/PH) in rats. We depleted macrophages in the liver of 2-AAF/PH treated rats by injecting liposome encapsulated clodronate 48 hours before PH. Regeneration of remnant liver mass, as well as proliferation and differentiation of oval cells were measured. We found that macrophage-depleted rats suffered higher mortality and liver transaminase levels. We also showed that depletion of macrophages yielded a significant decrease of EPCAM and PCK positive oval cells in immunohistochemical stained liver sections 9 days after PH. Meanwhile, oval cell differentiation was also attenuated as a result of macrophage depletion, as large foci of small basophilic hepatocytes were observed by day 9 following hepatectomy in control rats whereas they were almost absent in macrophage depleted rats. Accordingly, real-time polymerase chain reaction analysis showed lower expression of albumin mRNA in macrophage depleted livers. Then we assessed whether macrophage depletion may affect hepatic production of stimulating cytokines for liver regeneration. We showed that macrophage-depletion significantly inhibited hepatic expression of tumor necrosis factor-α and interleukin-6, along with a lack of signal transducer and activator of transcription 3 phosphorylation during the early period following hepatectomy. These data indicate that macrophages play an important role in oval cell mediated liver regeneration in the 2-AAF/PH model.
Sigma-Aldrich
N-(2-Fluorenyl)acetamide, ≥98% (HPLC)