Merck
  • beta-Lactam resistance in salmonella strains isolated from retail meats in the United States by the National Antimicrobial Resistance Monitoring System between 2002 and 2006.
beta-Lactam resistance in salmonella strains isolated from retail meats in the United States by the National Antimicrobial Resistance Monitoring System between 2002 and 2006.
Applied and environmental microbiology (2009-10-27)
S Zhao, K Blickenstaff, A Glenn, S L Ayers, S L Friedman, J W Abbott, P F McDermott
ABSTRACT
Ampicillin-resistant (Amp(r)) Salmonella enterica isolates (n = 344) representing 32 serotypes isolated from retail meats from 2002 to 2006 were tested for susceptibility to 21 other antimicrobial agents and screened for the presence of five beta-lactamase gene families (bla(CMY), bla(TEM), bla(SHV), bla(OXA), and bla(CTX-M)) and class 1 integrons. Among the Amp(r) isolates, 66.9% were resistant to five or more antimicrobials and 4.9% were resistant to 10 or more antimicrobials. Coresistance to other beta-lactams was noted for amoxicillin-clavulanic acid (55.5%), ceftiofur (50%), cefoxitin (50%), and ceftazidime (24.7%), whereas less than 5% of isolates were resistant to piperacillin-tazobactam (4.9%), cefotaxime (3.5%), ceftriaxone (2%), and aztreonam (1.2%). All isolates were susceptible to cefepime, imipenem, and cefquinome. No Salmonella producing extended-spectrum beta-lactamases was found in this study. Approximately 7% of the isolates displayed a typical multidrug-resistant (MDR)-AmpC phenotype, with resistance to ampicillin, chloramphenicol, streptomycin, sulfonamide, tetracycline, plus resistance to amoxicillin-clavulanic acid, cefoxitin, and ceftiofur and with decreased susceptibility to ceftriaxone (MIC > or = 4 microg/ml). Pulsed-field gel electrophoresis results showed that several MDR clones were geographically dispersed in different types of meats throughout the five sampling years. Additionally, 50% of the isolates contained bla(CMY), 47% carried bla(TEM-1), and 2.6% carried both genes. Only 15% of the isolates harbored class I integrons carrying various combinations of aadA, aadB, and dfrA gene cassettes. The bla(CMY), bla(TEM), and class 1 integrons were transferable through conjugation and/or transformation. Our findings indicate that a varied spectrum of coresistance traits is present in Amp(r) Salmonella strains in the meat supply of the United States, with a continued predominance of bla(CMY) and bla(TEM) genes in beta-lactam-resistant isolates.
Sigma-Aldrich
SOC Medium, For use in transformation
Pricing and availability is not currently available.