Merck
  • Siglec-6 as a therapeutic target for cell migration and adhesion in chronic lymphocytic leukemia.
Siglec-6 as a therapeutic target for cell migration and adhesion in chronic lymphocytic leukemia.
Nature communications (2024-06-19)
Jessica Nunes, Rakeb Tafesse, Charlene Mao, Matthew Purcell, Xiaokui Mo, Liwen Zhang, Meixiao Long, Matthew G Cyr, Christoph Rader, Natarajan Muthusamy
ABSTRACT
Siglec-6 is a lectin receptor with restricted expression in the placenta, mast cells and memory B-cells. Although Siglec-6 is expressed in patients with chronic lymphocytic leukemia (CLL), its pathophysiological role has not been elucidated. We describe here a role for Siglec-6 in migration and adhesion of CLL B cells to CLL- bone marrow stromal cells (BMSCs) in vitro and compromised migration to bone marrow and spleen in vivo. Mass spectrometry analysis revealed interaction of Siglec-6 with DOCK8, a guanine nucleotide exchange factor. Stimulation of MEC1-002 CLL cells with a Siglec-6 ligand, sTn, results in Cdc42 activation, WASP protein recruitment and F-actin polymerization, which are all associated with cell migration. Therapeutically, a Siglec-6/CD3-bispecific T-cell-recruiting antibody (T-biAb) improves overall survival in an immunocompetent mouse model and eliminates CLL cells in a patient derived xenograft model. Our findings thus reveal a migratory role for Siglec-6 in CLL, which can be therapeutically targeted using a Siglec-6 specific T-biAb.