Merck
  • Antinociceptive effect of intermittent fasting via the orexin pathway on formalin-induced acute pain in mice.
Antinociceptive effect of intermittent fasting via the orexin pathway on formalin-induced acute pain in mice.
Scientific reports (2023-11-21)
Hyunjin Shin, Jaehyuk Kim, Sheu-Ran Choi, Dong-Wook Kang, Ji-Young Moon, Dae-Hyun Roh, Miok Bae, Jungmo Hwang, Hyun-Woo Kim
ABSTRACT
It has been suggested that stress responses induced by fasting have analgesic effects on nociception by elevating the levels of stress-related hormones, while there is limited understanding of pain control mechanisms. Here, we investigated whether acute or intermittent fasting alleviates formalin-induced pain in mice and whether spinal orexin A (OXA) plays a role in this process. 6, 12, or 24 h acute fasting (AF) and 12 or 24 h intermittent fasting (IF) decreased the second phase of pain after intraplantar formalin administration. There was no difference in walking time in the rota-rod test and distance traveld in the open field test in all groups. Plasma corticosterone level and immobility time in the forced swim test were increased after 12 h AF, but not after 12 h IF. 12 h AF and IF increased not only the activation of OXA neurons in the lateral hypothalamus but also the expression of OXA in the lateral hypothalamus and spinal cord. Blockade of spinal orexin 1 receptor with SB334867 restored formalin-induced pain and spinal c-Fos immunoreactivity that were decreased after 12 h IF. These results suggest that 12 h IF produces antinociceptive effects on formalin-induced pain not by corticosterone elevation but by OXA-mediated pathway.