Merck
  • Molecular portraits of cell cycle checkpoint kinases in cancer evolution, progression, and treatment responsiveness.
Molecular portraits of cell cycle checkpoint kinases in cancer evolution, progression, and treatment responsiveness.
Science advances (2023-06-30)
Elena Oropeza, Sinem Seker, Sabrina Carrel, Aloran Mazumder, Daniel Lozano, Athena Jimenez, Sabrina N VandenHeuvel, Dillon A Noltensmeyer, Nindo B Punturi, Jonathan T Lei, Bora Lim, Susan E Waltz, Shreya A Raghavan, Matthew N Bainbridge, Svasti Haricharan
ABSTRACT
Cell cycle dysregulation is prerequisite for cancer formation. However, it is unknown whether the mode of dysregulation affects disease characteristics. Here, we conduct comprehensive analyses of cell cycle checkpoint dysregulation using patient data and experimental investigations. We find that ATM mutation predisposes the diagnosis of primary estrogen receptor (ER)+/human epidermal growth factor (HER)2- cancer in older women. Conversely, CHK2 dysregulation induces formation of metastatic, premenopausal ER+/HER2- breast cancer (P = 0.001) that is treatment-resistant (HR = 6.15, P = 0.01). Lastly, while mutations in ATR alone are rare, ATR/TP53 co-mutation is 12-fold enriched over expected in ER+/HER2- disease (P = 0.002) and associates with metastatic progression (HR = 2.01, P = 0.006). Concordantly, ATR dysregulation induces metastatic phenotypes in TP53 mutant, not wild-type, cells. Overall, we identify mode of cell cycle dysregulation as a distinct event that determines subtype, metastatic potential, and treatment responsiveness, providing rationale for reconsidering diagnostic classification through the lens of the mode of cell cycle dysregulation..
Sigma-Aldrich
Chk2 Inhibitor II hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.