Merck
  • Analysis of CF patient survival confirms STAT3 as a CF-modifying gene with changing impact over time.
Analysis of CF patient survival confirms STAT3 as a CF-modifying gene with changing impact over time.
Human molecular genetics (2022-09-02)
Inga Dunsche, Ellen L Raddatz, Haide Ismer, Silke Hedtfeld, Stephanie Tamm, Saskia Moser, Julia Kontsendorn, Burkhard Tümmler, Sabina Janciauskiene, Anna-Maria Dittrich, Frauke Stanke
ABSTRACT
The signal transducer and activator of transcription 3 (STAT3) has been identified as one of the cystic fibrosis (CF) modifying genes. In this study, we aimed to assess the association between STAT3 genotype and CF patient survival over several decades and to investigate the effect of STAT3 inhibition on epithelial CFTR expression. We analyzed the informative genetic marker STAT3Sat for its association with survival in 174 p.Phe508del-CFTR homozygous CF patients treated at the CF center in Hannover spanning birth cohorts from >3 decades (1959-1994). Furthermore, we treated two epithelial cell lines with STAT3 inhibitors and monitored changes of CFTR protein expression by western blot. Only for p.Phe508del-CFTR homozygous patients born prior to 1975, survival was significantly influenced by STAT3sat genotype (P = 0.023). The expression levels of STAT3 and CFTR positively correlated in epithelial cell lines (P = 0.01). Our results in different birth cohorts identified a time-dependent impact of STAT3 genotype on CF patients' survival and found that improved symptomatic treatment of later-born CF patients obviates STAT3's modifying influence. Consistent with our previous results, STAT3-specific inhibition resulted in increased CFTR expression in the epithelial cell line 16HBE14o-. Thus, care should be taken when CF-modifying genes are studied in cross-sectional cohorts as the impact of modifying genes might not be invariant in the light of changing therapeutic regimens.