Merck
  • Development of a COX-2-Selective Fluorescent Probe for the Observation of Early Intervertebral Disc Degeneration.
Development of a COX-2-Selective Fluorescent Probe for the Observation of Early Intervertebral Disc Degeneration.
Journal of functional biomaterials (2023-04-27)
Cheol Ho Heo, Eun Ji Roh, Jaehee Kim, Hyemin Choi, Ho Yeon Jang, Giseong Lee, Chang Su Lim, Inbo Han
ABSTRACT
Cyclooxygenase-2 (COX-2) is a biomolecule known to be overexpressed in inflammation. Therefore, it has been considered a diagnostically useful marker in numerous studies. In this study, we attempted to assess the correlation between COX-2 expression and the severity of intervertebral disc (IVD) degeneration using a COX-2-targeting fluorescent molecular compound that had not been extensively studied. This compound, indomethacin-adopted benzothiazole-pyranocarbazole (IBPC1), was synthesized by introducing indomethacin-a compound with known selectivity for COX-2-into a phosphor with a benzothiazole-pyranocarbazole structure. IBPC1 exhibited relatively high fluorescence intensity in cells pretreated with lipopolysaccharide, which induces inflammation. Furthermore, we observed significantly higher fluorescence in tissues with artificially damaged discs (modeling IVD degeneration) compared to normal disc tissues. These findings indicate that IBPC1 can meaningfully contribute to the study of the mechanism of IVD degeneration in living cells and tissues and to the development of therapeutic agents.
Sigma-Aldrich
COX-2 Inhibitor I, The COX-2 Inhibitor I, also referenced under CAS 416901-58-1, controls the biological activity of COX-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.