Merck
  • Nucleoredoxin interaction with flightless-I/actin complex is differentially altered in alcoholic liver disease.
Nucleoredoxin interaction with flightless-I/actin complex is differentially altered in alcoholic liver disease.
Basic & clinical pharmacology & toxicology (2020-06-12)
Brisa Rodope Alarcón-Sánchez, Dafne Guerrero-Escalera, Sandra Rosas-Madrigal, Diana Ivette Aparicio-Bautista, Karina Reyes-Gordillo, M Raj Lakshman, Arturo Ortiz-Fernández, Héctor Quezada, Óscar Medina-Contreras, Saúl Villa-Treviño, Julio Isael Pérez-Carreón, Jaime Arellanes-Robledo
ABSTRACT
Alcoholic liver disease (ALD) may be attributed to multiple hits driving several alterations. The aim of this work was to determine whether nucleoredoxin (NXN) interacts with flightless-I (FLII)/actin complex and how this ternary complex is altered during ALD progression induced by different ALD models. ALD was recapitulated in C57BL/6J female mice by the well-known ALD Lieber-DeCarli model, and by an in vitro human co-culture system overexpressing NXN. The effects of ethanol and low doses of lipopolysaccharides (LPS) and diethylnitrosamine (DEN) were also evaluated in vivo as a first approach of an ALD multi-hit protocol. We demonstrated that NXN interacts with FLII/actin complex. This complex was differentially altered in ALD in vivo and in vitro, and NXN overexpression partially reverted this alteration. We also showed that ethanol, LPS and DEN synergistically induced liver structural disarrangement, steatosis and inflammatory infiltration accompanied by increased levels of proliferation (Ki67), ethanol metabolism (CYP2E1), hepatocarcinogenesis (GSTP1) and LPS-inducible (MYD88 and TLR4) markers. In summary, we provide evidence showing that NXN/FLII/actin complex is involved in ALD progression and that NXN might be involved in the regulation of FLII/actin-dependent cellular functions. Moreover, we present a promising first approach of a multi-hit protocol to better recapitulate ALD pathogenesis.
Sigma-Aldrich
Anti-GSTP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution