Merck
  • Proteins associated with the Arabidopsis thaliana plastid rhomboid-like protein RBL10.
Proteins associated with the Arabidopsis thaliana plastid rhomboid-like protein RBL10.
The Plant journal : for cell and molecular biology (2021-09-29)
Anastasiya Lavell, Montgomery Smith, Yang Xu, John E Froehlich, Cameron De La Mora, Christoph Benning
ABSTRACT
Rhomboid-like proteins are intramembrane proteases with a variety of regulatory roles in cells. Though many rhomboid-like proteins are predicted in plants, their detailed molecular mechanisms or cellular functions are not yet known. Of the 13 predicted rhomboids in Arabidopsis thaliana, one, RBL10, affects lipid metabolism in the chloroplast, because in the respective rbl10 mutant the transfer of phosphatidic acid through the inner envelope membrane is disrupted. Here we show that RBL10 is part of a high-molecular-weight complex of 250 kDa or greater in size. Nine likely components of this complex are identified by two independent methods and include Acyl Carrier Protein 4 (ACP4) and Carboxyltransferase Interactor1 (CTI1), which have known roles in chloroplast lipid metabolism. The acp4 mutant has decreased C16:3 fatty acid content of monogalactosyldiacylglycerol, similar to the rbl10 mutant, prompting us to offer a mechanistic model of how an interaction between ACP4 and RBL10 might affect chloroplast lipid assembly. We also demonstrate the presence of a seventh transmembrane domain in RBL10, refining the currently accepted topology of this protein. Taken together, the identity of possible RBL10 complex components as well as insights into RBL10 topology and distribution in the membrane provide a stepping-stone towards a deeper understanding of RBL10 function in Arabidopsis lipid metabolism.
Product Description
Supelco
Protein Standard Mix 15 - 600 kDa, for size exclusion chromatography
Pricing and availability is not currently available.