Merck
  • Large-scale isolation of functional dermal papilla cells using novel surface marker LEPTIN Receptor.
Large-scale isolation of functional dermal papilla cells using novel surface marker LEPTIN Receptor.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2022-05-08)
Lipeng Gao, Eve Qian Chen, Hong-Bing Zhong, Jing Xie, Hong-Zhi Song, Xu-Bo Zhao, Jin-Ran Lin, Qingmei Liu, Shihua Wang, Wen-Yu Wu, Robert Chunhua Zhao, Xin-Hua Liao
ABSTRACT
Dermal papilla (DP) cells regulate hair follicle epithelial cells and melanocytes by secreting functional factors, playing a key role in hair follicle morphogenesis and hair growth. DP cells can reconstitute new hair follicles and induce hair regeneration, providing a potential therapeutic strategy for treating hair loss. However, current methods for isolating DP cells are either inefficient (physical microdissection) or only applied to genetically labeled mice. We systematically screened for the surface proteins specifically expressed in skin DP using mRNA expression databases. We identified two antibodies against receptors LEPTIN Receptor (LEPR ) and Scavenger Receptor Class A Member 5 (SCARA5) which could specifically label and isolate DP cells by flow cytometry from mice back skin at the growth phase. The sorted LEPR+ cells maintained the DP characteristics after culturing in vitro, expressing DP marker alkaline phosphatase and functional factors including RSPO1/2 and EDN3, the three major DP secretory factors that regulate hair follicle epithelial cells and melanocytes. Furthermore, the low-passage LEPR+ DP cells could reconstitute hair follicles on nude mice using chamber graft assay when combined with epithelial stem cells. The method of isolating functional DP cells we established here lays a solid foundation for developing DP cell-based therapy.