Merck
  • G protein antagonists. A novel hydrophobic peptide competes with receptor for G protein binding.
G protein antagonists. A novel hydrophobic peptide competes with receptor for G protein binding.
The Journal of biological chemistry (1992-08-15)
H Mukai, E Munekata, T Higashijima
ABSTRACT
A substance P (SP) analog, [D-Pro4,D-Trp7,9,10] SP4-11, is known to inhibit the actions of various structurally unrelated messenger molecules as well as SP. Our studies on the effects of this peptide on the regulation of purified G proteins by receptor showed that at least some of the biological effects of the peptide can be explained by the ability of the peptide to block the activation of G proteins by receptors. Here we report that a novel truncated SP-related peptide, pGlu-Gln-D-Trp-Phe-D-Trp-D-Trp-Met-NH2, inhibited the activation of G(i) or G(o) by M2 muscarinic cholinergic receptor (M2 mAChR) or of Gs by beta-adrenergic receptor in the reconstituted phospholipid vesicles, assayed by receptor-promoted GTP hydrolysis. The inhibition by the peptide was apparently reversible and competitive with respect to receptor binding to G proteins; the inhibition could be overcome by increasing the concentration of receptor in the vesicles and was not altered by changes in the concentration of G protein. The competing effects of the peptide were used to analyze the effect of agonist on receptor-G protein interaction. The concentration change of muscarinic agonist did not alter the inhibitory effects of the peptide on M2 mAChR-promoted GTPase by G(o), which is consistent with the idea that agonist increases the regulatory efficiency of the receptor but does not alter its affinity for G proteins. This new group of compounds (G protein antagonists) is a promising tool to study receptor-G protein interaction quantitatively.
Sigma-Aldrich
[pGlu5, D-Trp7,9,10]-Substance P Fragment 5-11, ≥90% (HPLC)