Merck
  • RHOA protein expression correlates with clinical features in gastric cancer: a systematic review and meta-analysis.
RHOA protein expression correlates with clinical features in gastric cancer: a systematic review and meta-analysis.
BMC cancer (2022-07-20)
Seungyoon Nam, Yeeun Lee, Jung Ho Kim
ABSTRACT
Gastric cancer (GC) is one of the most fatal cancers worldwide and is generally only detected after it has progressed to an advanced stage. Since there is a lack of comprehensive data on RHOA protein expression of patients with GC, this study utilized a systematic review and meta-analysis to address the limitation. The objective of this meta-analysis was to link GC clinical features with RHOA protein high- vs. low-expressing patients with GC. The PubMed and Web of Science were used for a systematic literature review of GC related to RHOA. The included studies were obtained from two literature databases from past to Aug 31, 2021, by searching keywords. This meta-analysis followed the Preferred Reporting Items for Systematic Reviews and Meta-Analysis (PRISMA) guidelines. The odds ratios (ORs) and 95% confidential intervals (CIs) for clinical features were estimated according to the high and low protein expression levels of RhoA. The mean effect sizes of ORs were obtained using the random-effects and fixed-effects models of meta-analysis. Heterogeneity of the studies was assesed by using statistics: τ2, I2; and Q values. The symmetry of funnel plots were inspected for publication bias. Finally, 10 studies including 1,389 patients with GC (735 RHOA-positive and 654 RHOA-negative) were eligible for our meta-analysis to estimate associations between the protein expression and clinical features (e.g., Union for International Cancer Control [UICC] stage progression, differentiation, Lauren histological classification, and vascular invasion). In our meta-analysis, RHOA positive expression was determined to have a statistically significant association with UICC stage progression (P = 0.02) and poorly differentiated status (P = 0.02). The association between RHOA positivity and Lauren subtypes was not statistically significant (P = 0.07). This meta-analysis suggested that RhoA protein expression in patients with GC was associated with clinical features: UICC stage progression and poorly differentiated status. Our findings are inconclusive but indicate that high RHOA protein expressing patients with GC could predict advanced UICC stages. A large prospective cohort study is required for validation in future.