Merck
  • Predicting the Orientation of Adsorbed Proteins Steered with Electric Fields Using a Simple Electrostatic Model.
Predicting the Orientation of Adsorbed Proteins Steered with Electric Fields Using a Simple Electrostatic Model.
The journal of physical chemistry. B (2022-07-13)
Sergio A Urzúa, Perla Y Sauceda-Oloño, Carlos D García, Christopher D Cooper
ABSTRACT
Under the most common experimental conditions, the adsorption of proteins to solid surfaces is a spontaneous process that leads to a rather compact layer of randomly oriented molecules. However, controlling such orientation is critically important for the development of catalytic surfaces. In this regard, the use of electric fields is one of the most promising alternatives. Our work is motivated by experimental observations that show important differences in catalytic activity of a trypsin-covered surface, which depended on the applied potential during the adsorption. Even though adsorption results from the combination of several processes, we were able to determine that (under the selected conditions) mean-field electrostatics play a dominant role, determining the orientation and yielding a difference in catalytic activity. We simulated the electrostatic potential numerically, using an implicit-solvent model based on the linearized Poisson-Boltzmann equation. This was implemented in an extension of the code PyGBe that included an external electric field, and rendered the electrostatic component of the solvation free energy. Our model (extensions available at the Github repository) allowed estimating the overall affinity of the protein with the surface, and their most likely orientation as a function of the potential applied. Our results show that the active sites of trypsin are, on average, more exposed when the electric field is negative, which agrees with the experimental results of catalytic activity, and confirm the premise that electrostatic interactions can be used to control the orientation of adsorbed proteins.
Sigma-Aldrich
Trypsin Activity Colorimetric Assay Kit, sufficient for 100 colorimetric tests