Merck
Fungal phytochrome chromophore biosynthesis at mitochondria.
The EMBO journal (2021-07-14)
Christian Streng, Jana Hartmann, Kai Leister, Norbert Krauß, Tilman Lamparter, Nicole Frankenberg-Dinkel, Franco Weth, Martin Bastmeyer, Zhenzhong Yu, Reinhard Fischer
ABSTRACT
Mitochondria are essential organelles because of their function in energy conservation. Here, we show an involvement of mitochondria in phytochrome-dependent light sensing in fungi. Phytochrome photoreceptors are found in plants, bacteria, and fungi and contain a linear, heme-derived tetrapyrrole as chromophore. Linearization of heme requires heme oxygenases (HOs) which reside inside chloroplasts in planta. Despite the poor degree of conservation of HOs, we identified two candidates in the fungus Alternaria alternata. Deletion of either one phenocopied phytochrome deletion. The two enzymes had a cooperative effect and physically interacted with phytochrome, suggesting metabolon formation. The metabolon was attached to the surface of mitochondria with a C-terminal anchor (CTA) sequence in HoxA. The CTA was necessary and sufficient for mitochondrial targeting. The affinity of phytochrome apoprotein to HoxA was 57,000-fold higher than the affinity of the holoprotein, suggesting a "kiss-and-go" mechanism for chromophore loading and a function of mitochondria as assembly platforms for functional phytochrome. Hence, two alternative approaches for chromophore biosynthesis and insertion into phytochrome evolved in plants and fungi.
Sigma-Aldrich
Yeast Mitochondria Isolation Kit, sufficient for 40 applications (using 20 OD culture preparations), isolation of an enriched mitochondrial fraction from yeast cells